Would you like to log in again?
CrossLab Gas Cylinder Supplies, Agilent Technologies
Supplier: Agilent
CrossLab
0460-1266 19246-60570 5180-4143 19246-60690 0460-1266 0100-0119 0100-0110 0100-0110 5183-4642 0101-1535 5183-4645 G4218-60100 5188-5374 0100-0119 5183-1941 19246-60560 19246-60570 5183-4641 5188-5374 5180-4145 0100-0111 5183-4644 0100-0118 0101-1535 5180-4143 5183-4645 5183-4641 0101-1400 5183-4643 5183-4644 5183-4643 5183-4642 0100-0118 G4218-60100 0100-0111 19246-60690 0101-1536 5183-1941 19246-60560 0101-1536 5180-4145
AG19246-60690EA 364.86 USD InStock
CAAG0100-0110 AG19246-60690 AG19246-60570 CA0100-0119 AG0100-0110 CA0100-0118 AG0100-0111 CA5183-1941 CAAGG421860100 AG0100-0118 AG0100-0119 CAAG5188-5374 CAAG1924660690 CAAG1924660570 AG5188-5374 CA0100-0111 AG5183-1941 AG0101-1400 CA5183-4645 AG19246-60560 AG0460-1266 CAAG5180-4145 AG5180-4143 CAAG5183-4643 CA5183-4642 AG5183-4641 CA5180-4143 CA5183-4641 CA0460-1266 CA76514-896 CAAG0101-1535 CAAG0101-1536 CAAG1924660560 AG5183-4645 AG5183-4644 AG5183-4643 AG5183-4642 AG0101-1536 AG0101-1535 AG5180-4145 AGG4218-60100
CrossLab Gas Cylinder Supplies, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
Agilent gas cylinder supplies include economical brass regulators, wall brackets, and NPT adapters.
  • Dual-stage regulators provide more precise and consistent pressure control than single-stage regulators;Maximum inlet pressure: 3000 psig
  • Operating temperature range: ‒40 to 165 °F
  • Swagelok regulator for female flare NPT adapter available in ¹/₈ or ¹/₄ brass connector
Agilent gas cylinder supplies include regulators that provide proper gas pressure control for gas chromatography. These brass-body regulators are available for US only and follow the Compressed Gas Association (CGA) connection guidelines. Each gas regulator comes with ¹/₈ inch pipe thread fitting.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty