Would you like to log in again?
CrossLab Gas Chromatography (GC) Interface Supplies for ICP-MS, Agilent Technologies
Supplier: Agilent Technologies
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
CrossLab
0100-1331 G3158-80081 G3158-80150 G3158-80074 G3158-60010 G3158-80150 0100-1331 G3158-80060 G3158-80074 G3158-80060 G3158-80081 G3158-60010 G3158-80080 G3158-80080
AGG3158-80081EA 560 USD InStock
AGG3158-80081 AG0100-1331 AGG3158-80060 AGG3158-80080 CAAGG315880150 AGG3158-80074 CAAGG315880074 AGG3158-80150 AGG3158-60010 CAAGG315860010 CAAGG315880080 CAAGG315880081 CAAGG315880060 CA0100-1331
CrossLab Gas Chromatography (GC) Interface Supplies for ICP-MS, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
Agilent is an expert in speciation analysis, offering GC-ICP-MS parts and interface supplies.
  • The Agilent GC-ICP-MS interface consists of a heated, passivated transfer line and a special torch with a heated injector tube
  • The unique interface is ideal for routine analysis of high-boiling point compounds, providing unparalleled performance
  • All GC-ICP-MS interface supplies are designed to Agilent’s demanding specifications and can help to achieve the market-leading ICP-MS performance that our valued clients have come to expect
  • The GC interface function is enabled with PFA tubes, transfer lines and torches for the specific interface along with stainless pipe (Sulfinert) and clamp plates (PEEK) for straightforward installation and maintenance
  • Argon make-up gas supplied by the ICP-MS is preheated via a stainless steel heat exchanger inside the GC oven before being added to the GC column outlet to maintain constant high temperature from the end of the GC column to the tip of the ICP injector
The Agilent GC-ICP-MS interface consists of a heated, passivated transfer line and a special torch with a heated injector tube. The unique GC-ICP-MS interface is ideal for routine analysis of high-boiling point compounds, providing unparalleled performance for advanced GC-ICP-MS applications.
Argon make-up gas supplied by the ICP-MS is preheated via a stainless steel heat exchanger inside the GC oven before being added to the GC column effluent at the head of the transfer line. In this way, the sample is maintained at constant high temperature from the end of the chromatographic column in the GC oven to the tip of the ICP injector.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty