Would you like to log in again?
CrossLab Adapters and Unions for HPLC, Agilent Technologies
Supplier: Agilent
5067-6127 5022-2133 5042-8518 0100-1847 5022-2144 5022-2144 5067-4741 5065-9971 0100-2298 0100-2441 0100-0900 5022-2133 0100-2441 0100-0900 5022-2145 5067-4741 0100-1847 0100-2298 5042-8517 5022-2184 5065-9971 5022-2145 5022-2184 5042-8518 5067-6127 5042-8517
AG5022-2133EA 66.79 USD InStock
AG5022-2133 CAAG5065-9971 AG0100-2298 CA76514-622 CA5022-2133 AG5042-8517 AG5065-9971 AG5042-8518 CA5067-6127 CA5067-4741 CA76514-496 AG0100-1847 CA0100-2298 AG5022-2145 CA0100-0900 AG5022-2144 AG0100-2441 AG5022-2184 CA5022-2145 AG0100-0900 CAAG5042-8517 CAAG5042-8518 CAAG5022-2144 CA0100-2441 AG5067-6127 AG5067-4741
CrossLab Adapters and Unions for HPLC, Agilent Technologies
Fittings and Connectors
High-quality LC unions and adapters for a range of equipment and applications.
  • Agilent quality parts are designed, tested, and manufactured with the same attention to detail you expect from Agilent instruments
  • Using non-Agilent supplies can considerably shorten lifetime and increases potential baseline noise
Agilent manufacture a wide variety of high-quality LC Unions suitable for a range of equipment and applications, including stainless steel ZDV unions (with and without fittings) for standard LC, ZDV unions (PEEK or stainless steel with PEEK inserts) for bio applications, and high-flow unions (no fittings) for prep LC. A range of adapters and connectors are also available, including barbed Y-connectors, T-connectors, and female-to-female or male-to-female adapters.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty