Maintenance kits with all the necessary parts for keeping your HPLC instrument at peak performance.
- Lack of preventive maintenance can result in noisy baselines, shorter column life, lower analytical sensitivity, and variable retention times
- Routine maintenance should be done on a regular basis to keep your Agilent LC system performing at its optimum
- Agilent preventive maintenance kits ensure HPLC systems are at their best
- Maintenance procedures can be performed all at once or as needed
Agilent provides a full range of HPLC maintenance kits that include all supplies to be exchanged on a regular basis to ensure the instrument is always operating at peak performance. All parts are manufactured to Agilent's highest levels of quality and have been tested in Agilent instruments to ensure they work perfectly with the rest of the system.