Would you like to log in again?
Cell Holders, Bases & Accessories for UV-Vis & UV-Vis-NIR, Agilent Technologies
Supplier: Agilent Technologies
CrossLab
7910047600 210125300 08451-60104 110059900 7910033500 7910038700 210125300 7910047600 110059900 7910033500 08451-60104 7910038700
AG210125300KT 833 USD InStock
CAAG110059900 CAAG210125300 AG210125300 AG110059900 CAAG7910038700 CAAG7910047600 AG7910047600 CAAG7910033500 AG08451-60104 AG7910038700 AG7910033500 CA08451-60104
Cell Holders, Bases & Accessories for UV-Vis & UV-Vis-NIR, Agilent Technologies
Spectrophotometers and Photometers UV/Visible Spectrophotometers
Standard, cylindrical, long and variable pathlength cell holders and bases for Agilent UV-Vis systems.
  • Standard cell holders replace those supplied with the instrument
  • Standard cell holder ensures quick and precise cell positioning for each measurement
  • Standard cell holders include cell lifter and adjustment screw
  • Long pathlength cell holders have fixed mounting positions that enable rectangular cells up to 100 mm pathlength to be used
  • Variable pathlength cell holders enable rectangular cells with pathlengths from 5 to 50 mm to be used
  • Cylindrical cell holder allows use of cylindrical cells with up to 100 mm pathlength
  • Test tube holder allows use of 16 mm od test tubes in the sample compartment instead of conventional cuvettes
Agilent offers a range of cell holders, providing you with the ability to analyze a wide variety of samples. Standard, cylindrical, long pathlength, and variable pathlength cell holders, bases and test tube holders are all available for Agilent UV-Vis and UV-Vis-NIR systems.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty