Would you like to log in again?
Liner O-Rings for GC, Agilent Technologies
Supplier: Agilent Technologies
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
CrossLab
5188-5365 MKI-U-COV10 0905-1145 5180-4168 5061-5867 0905-1608 5190-2268 MKI-U-COV10 5180-4181 5190-2268 5188-5311 5180-4168 5080-4982 5061-5867 0905-1609 5188-5311 5180-4173 0905-1608 0905-1610 0905-1609 5190-2269 5080-4982 0905-1145 5180-4181 5180-4173 0905-1610 5190-2269 5188-5365
AG5180-4173PK 86.7 USD InStock
AG5180-4173 AG5061-5867 AG5190-2269 AG5190-2268 CA0905-1145 AGMKI-U-COV10 CA5180-4173 CAMKI-U-COV10 AG0905-1145 CAAG5188-5311 AG5188-5311 AG5180-4181 CAAG0905-1610 CA5180-4168 CA5080-4982 AG0905-1610 CA5180-4181 CA76508-184 CAAG0905-1608 AG0905-1608 AG0905-1609 CAAG0905-1609 AG5080-4982 CA5061-5867 AG5188-5365 CA5190-2268 CA5190-2269 AG5180-4168
Liner O-Rings for GC, Agilent Technologies
Seals Sealing Rings
Liner O-rings for GC and GC/MS systems.
  • Liners are sealed in the inlet with O-rings or graphite seals
  • Graphite seals are used when inlet temperatures exceed 350 °C
  • Fluorocarbon O-ring seals are easier to replace than graphite that deforms and flakes apart
  • Plasma treated for a nonstick, contaminant-free surface that won’t stick to the inlet metal surface
  • Packaged for convenience and cleanliness in a novel dial package that delivers 1 clean O-ring at a time.
Liner O-rings are used to form a proper seal around the outside of the inlet liner between the inlet weldment and the turn-top. O-rings are composed of either graphite or fluorocarbon material, graphite O-rings are used when operating inlet temperatures exceed 350 °C.
Only Agilent Fluorocarbon Liner O-rings are Precleaned, then conditioned to eliminate out-gassing of contaminants, which is especially important for trace, ECD and MSD analyses.
Packaging: Packaged for convenience and cleanliness in a novel dial package that delivers one clean O-ring at a time.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty