Would you like to log in again?
CrossLab CAM Columns, Agilent Technologies
Supplier: Agilent Technologies
CrossLab
113-2132 112-2132 113-2132 112-2132
AG112-2132EA 787 USD InStock
CA112-2132 AG112-2132 AG113-2132 CA113-2132
CrossLab CAM Columns, Agilent Technologies
Chromatography Columns
A base-deactivated PEG column specifically designed for amine analysis, delivering excellent peak shape for primary amines.
  • Base-deactivated polyethylene glycol
  • Specifically designed for amines analysis
  • Excellent peak shape for primary amines
  • Replaces HP-Basicwax
  • Tested with the tightest industry QC specifications for column bleed, sensitivity, and efficiency
  • Performance Summary shipped with each GC column
  • Not bonded or crosslinked, not recommended for solvent rinsing
Agilent J&W CAM is a polyethylene glycol column that delivers excellent peak shape for primary amines. CAM replaces HP-BasicWax. The column is not bonded or crosslinked, so solvent rinsing is not recommended.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty