Would you like to log in again?
GS-CarbonPLOT Columns, Agilent Technologies
Supplier: Agilent
CrossLab
115-3133 115-3133 113-3133 113-3133
AG115-3133EA 1059.26 USD InStock
CA115-3133 AG115-3133 CA113-3133 AG113-3133
GS-CarbonPLOT Columns, Agilent Technologies
Chromatography Columns
A high-stability, carbon-layer stationary phase with unique selectivity for inorganic and organic gases.
  • High-stability, carbon-layer stationary phase
  • Unique selectivity for inorganic and organic gases
  • Extended temperature limit of 360 °C
  • Ideal for GC/MS, due to no particle generation
  • Retention stability not affected by water
  • Tested with the tightest industry QC specifications for column bleed, sensitivity, and efficiency
  • Performance Summary shipped with each column
Agilent J&W GS-CarbonPLOT uses a high-stability, carbon-layer stationary phase to deliver unique selectivity for inorganic and organic gases. The column features an extended temperature limit to 360 °C. Retention stability is not affected by water, and the column is ideal for GC/MS because its bleed-free phase does not shed particles.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty