Would you like to log in again?
CrossLab Dissolution Baskets, Agilent Technologies
Supplier: Agilent
CrossLab
12-2154 12-2154 12-2146 12-2155 12-2155 12-2152 12-2152 12-2185 12-2100V 12-2185 12-2151 12-2151 12-2180 12-2146 12-2180 27-3007 12-2100V 27-3007
AG12-2100VEA 465.65 USD InStock
CAAG12-2146 AG12-2100V CA12-2100V AG12-2151 AG12-2152 AG12-2185 CAAG12-2151 AG12-2154 AG12-2155 CAAG12-2152 CAAG12-2185 CAAG12-2155 CAAG12-2154 AG12-2146 CAAG27-3007 AG27-3007 AG12-2180 CAAG12-2180
CrossLab Dissolution Baskets, Agilent Technologies
Dissolution Apparatus
USP 40-mesh stainless steel baskets, custom baskets, and baskets for small or large dosage forms.
  • A variety of basket and mesh sizes available
  • Available in different materials and coatings
  • Individual Certificate of Conformity can be provided
From the standard USP 40-mesh stainless steel basket, to custom baskets with varying mesh sizes, to baskets specific to small or large dosage forms, Agilent supports your needs. Unless otherwise noted, all baskets fit the standard basket shafts.
Certifications: Individual Certificate of Conformity can be provided.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty