Would you like to log in again?
Dissolution Sinkers, Agilent Technologies
Supplier: Agilent Technologies
12-3070 12-3000 12-3000 12-3070
AG12-3070EA 354 USD InStock
AG12-3070 CAAG12-3070 AG12-3000 CAAG12-3000
Dissolution Sinkers, Agilent Technologies
Dissolution Apparatus
A variety of dissolution sinkers are available depending on your dosage form.
  • Capsule wire, weights, and sinker baskets are used to retain a dosage form at the bottom of the vessel for testing when the dosage form is buoyant
  • A variety of options are available
  • The flexible capsule wire meets USP requirements and has a 0.032" diameter
  • Sinker baskets are offered in a variety of mesh sizes and made of stainless steel with press-on caps, using O-rings to seal the basket
  • Japanese Pharmacopoeia (JP) sinker baskets are available as an alternative
  • Three-prong capsule weights fit traditional capsule sizes one through three, and are sold individually
Capsule wire, weights, and sinker baskets are used to retain a solid dosage form at the bottom of the vessel for testing when the dosage form is buoyant. Depending on your dosage form, a variety of options are available.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty