Would you like to log in again?
CrossLab Dissolution Vessels and Carriers, Agilent Technologies
Supplier: Agilent
CrossLab
12-5157V 27-6050 12-5035 12-5149 12-5146 12-5149 12-5035V 27-6000 12-5010 27-5110 12-5147 12-5010V 12-5146 27-6050 12-5158 12-5158 12-5120 12-5070 12-5050 33-9005 12-5145 12-5125 12-5159 12-5120 33-9000 12-5000 12-5040 12-5033 12-5158V 12-5147 12-5055 12-5505 12-5149V 12-5152V 12-5157V 12-5035 12-5035V 12-5500 12-5505 12-5055 27-5130 12-5200 27-5005 12-5034 12-5000 12-5010 12-5153 27-5000 12-5000V 12-5157 27-5100 33-9005 12-5500 12-5159 12-5040 27-5000 13-0010 12-5153 12-5152V 12-5070 12-5050 12-5148 27-5130 12-5000V 12-5125 13-0010 12-5158V 12-5200 27-5100 12-5034 12-5033 27-6000 12-5010V 27-5120 12-5149V 27-5120 12-5157 12-5145 12-5148
AG27-5100EA 136.39 USD InStock
CAAG12-5500 CAAG12-5148 CAAG12-5149 AG27-5100 CAAG12-5505 CAAG27-6000 AG12-5010 AG12-5055 CAAG12-5146 AG12-5158V CAAG12-5147 AG33-9005 CAAG12-5145 CAAG12-5070 CAAG12-5152V CAAG13-0010 CAAG12-5035V AG12-5505 CAAG12-5159 AG27-5130 CAAG12-5153 AG12-5000V CAAG12-5033 AG12-5145 AG12-5500 AG12-5148 CAAG12-5157 AG12-5149 CAAG12-5158 AG12-5146 CAAG12-5034 AG12-5147 AG12-5157V CAAG12-5035 AG12-5070 AG12-5149V CAAG12-5040 AG27-5005 CAAG12-5157V CA27-5120 CA27-5000 CAAG12-5000V CAAG12-5158V CAAG27-5130 AG27-5120 AG27-5000 CAAG12-5120 AG12-5034 AG12-5035 CAAG12-5000 AG12-5153 AG12-5152V AG12-5033 AG12-5159 CAAG12-5125 AG27-6050 AG12-5035V AG12-5157 AG12-5158 CAAG12-5200 CAAG12-5050 AG12-5040 CAAG27-6050 AG27-6000 AG27-5110 CAAG12-5010 CAAG33-9005 CAAG12-5055 AG12-5200 AG12-5120 AG12-5000 AG12-5010V AG12-5125 CA12-5149V AG33-9000 AG12-5050 CAAG12-5010V AG13-0010 CA27-5100
CrossLab Dissolution Vessels and Carriers, Agilent Technologies
Dissolution Apparatus
EaseAlign, TruCenter and TruAlign dissolution vessels are available in a variety of sizes.
  • Vessel volume sizes include 100 and 200 ml, and 1 and 2 L sizes
  • Low actinic-red, peak, and verified versions are available for select volumes
  • Verified dissolution vessels and other accessories are available to support conformance guidelines for mechanical qualification
  • Conversion kits are available to support small- and large-volume dissolution testing
Agilent provides a variety of dissolution vessels like TruAlign, TruCenter, or EaseAlign vessels. These are available with or without a Certificate of Conformance (COC) for Agilent dissolution apparatus. Tightest precision is available with the vacuum-formed (VF) vessels.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty