Would you like to log in again?
J&W High Efficiency Columns, Agilent Technologies
Supplier: Agilent
CP7461 CP7411 121-1544 121-7022 121-1324 121-9627 121-9627 121-1544 121-9723 121-7022 121-1524 121-1324 CP7340 CP7686 CP7461 CP7340 121-1524 CP7411 CP7686 121-9723
AG121-9627EA 785.44 USD InStock
CA121-9627 AG121-9627 AG121-9723 AG121-1544 CA121-1324 CA121-1544 AG121-1324 AG121-1524 AGCP7411 AGCP7686 CA121-7022 CAAG121-9723 AGCP7340 AGCP7461 CA121-1524 CAAGCP7411 CAAGCP7686 CAAGCP7461 CAAGCP7340 AG121-7022
J&W High Efficiency Columns, Agilent Technologies
Chromatography Columns
A simple and cost-effective way to increase sample throughput with existing GC or GC/MS instruments.
  • Increase sample throughput and productivity while maintaining resolution
  • Reduce carrier gas usage and cost per analysis
  • Apply with existing GC and GC/MS instrument
  • Operates with He or H2 carrier gas
  • Available in more than 20 stationary phases
  • Rapid and easy method translation
  • Tested with the tightest industry QC specifications for column bleed, sensitivity, and efficiency
  • Performance Summary shipped with each column
Agilent High Efficiency GC columns are ideal for applications that require faster run times, such as high-throughput screening, fast process monitoring, and fast method development. They can reduce sample run time by 50% or more without compromising resolution.
Unlike 0.1 mm id columns, Agilent 0.15 and 0.18 mm id columns are compatible with all standard-pressure capillary GC and GC/MS instruments—without requiring expensive high-pressure modifications.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty