Would you like to log in again?
Bond Elut PH, Agilent Technologies
Supplier: Agilent
CrossLab
14102062 12102032 14102032 12102062 12102032 14256004 14113005 14113005 12102062 14102062 14256004 14102032
AG14102032PK 192.79 USD InStock
AG14102032 CAAG12102062 CAAG14102032 AG14102062 CAAG14102062 AG12102032 AG12102062 CAAG12102032 CAAG14256004 CAAG14113005 AG14113005 AG14256004
Bond Elut PH, Agilent Technologies
Chromatography Columns
Bond Elut PH is a moderately nonpolar sorbent commonly used for nonpolar extractions.
  • Added selectivity compared to other nonpolar sorbents
  • Enhanced retention of planar, conjugated organic molecules
  • Similar polarity to C8
Bond Elut PH is a nonpolar bonded silica material that exhibits a selectivity different from alkyl or aliphatic-functionalized phases, such as C8 or cyclohexyl. The electron density in the aromatic ring enhances retention of conjugated or aromatic ring-containing analytes due to desirable pi-pi interactions.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty