Would you like to log in again?
Bond Elut Certify II, Agilent Technologies
Supplier: Agilent Technologies
Bond Elut™
12113051 12113051 14102088 12102080 14102084 14113051 14102084 12102080 14102088 14113051
AG14102088PK 203 USD InStock
CAAG12102080 AG14102088 CAAG14102088 AG14113051 AG14102084 CAAG14113051 AG12102080 CAAG12113051 CAAG14102084 AG12113051
Bond Elut Certify II, Agilent Technologies
Chromatography Columns
Bond Elut Certify II is a SAX/nonpolar SPE phase for extracting acidic analytes from biomatrices.
  • Ideal for nonpolar and anionic compounds
  • Optimized for acidic drug analysis
  • Bimodal, nonpolar and strong anion exchange
Bond Elut Certify II was developed specifically for the rapid extraction of acidic drugs and metabolites from urine and other biological matrices. Certify II is a mixed-mode cartridge packed with nonpolar C8 and SAX sorbent. It has been optimized for acidic drugs and other compounds that possess both nonpolar and anionic characteristics.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty