Would you like to log in again?
Bond Elut Sodium Sulfate Drying Cartridges, Agilent Technologies
Supplier: Agilent Technologies
CrossLab
12132004 12131033 12131033 12162052B 12162052B 12132004
AG12162052BPK 412 USD InStock
CAAG12131033 AG12162052B CAAG12162052B AG12131033 CAAG12132004 AG12132004
Bond Elut Sodium Sulfate Drying Cartridges, Agilent Technologies
Chromatography Columns
Bond Elut sodium sulfate drying cartridges simplify drying processes with prepacked devices.
  • Easy to use
  • Available in different formats
  • Prepacked for convenience
Simplify sodium sulfate-mediated drying steps by using cartridges prepacked with ACS grade, granular, anhydrous sodium sulfate. Available in three formats: LRC (large reservoir capacity), Bond Elut Jr, and straight barrels.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty