Would you like to log in again?
J&W DB-XLB Columns, Agilent Technologies
Supplier: Agilent
J&W
121-1222 122-1232 122-1232 123-1232 123-1236 121-1222 123-1262 128-1222 123-1236-INT 122-1236 125-1212 122-1262 122-1262 123-1236-INT 123-1236 125-1232 123-1262 122-1236 123-1232 125-1212 125-1232
AG121-1222EA 696.9 USD InStock
AG121-1222 CA121-1222 AG123-1236-INT CAAG123-1262 AG123-1232 AG122-1262 AG123-1236 AG125-1232 AG128-1222 CAAG1231236INT AG125-1212 AG122-1236 CA122-1236 AG122-1232 CA122-1232 AG123-1262 CA122-1262 CA123-1232 CAAG125-1212 CAAG125-1232 CA123-1236
J&W DB-XLB Columns, Agilent Technologies
Chromatography Columns
Exceptionally low bleed for active compounds, ideal for GC/MS applications and confirmation analysis.
  • Exceptionally low bleed and low polarity
  • Unique selectivity
  • Excellent inertness for active compounds
  • Ideal for confirmation analysis
  • Excellent for pesticides, herbicides, PCBs, and PAHs
  • Well suited for GC/MS
  • MSD testing and certification available
  • Bonded, crosslinked, and solvent rinsable
  • Available with DuraGuard (GC column with built-in guard column)
  • Tested with the tightest industry QC specifications for column bleed, sensitivity, and efficiency
  • Performance Summary shipped with each column
  • Similar phases: Rtx-XLB, MDN-12, ZB-XLB, ZB-XLB HT
Agilent J&W DB-XLB has exceptionally low bleed and low polarity, and delivers highly specific analysis of pesticides, herbicides, polychlorinated biphenyls (PCBs), and polyaromatic hydrocarbons (PAHs).
DB-XLB is ideal for GC/MS applications and is also an excellent choice as a confirmation column for semivolatile environmental analysis.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty