Would you like to log in again?
J&W DB-1301 Columns, Agilent Technologies
Supplier: Agilent Technologies
J&W
122-1333 123-1333 125-1332 122-1333 125-1333 123-1333 125-1312 125-1312 123-1363 125-1333 122-1363 122-1363 123-1332 122-1332 122-1332 123-1332 123-1363 125-1332
AG123-1332EA 764 USD InStock
CAAG125-1312 CAAG122-1332 CAAG122-1333 CAAG123-1363 AG123-1332 AG123-1333 AG123-1363 AG122-1333 AG125-1312 AG125-1333 AG125-1332 AG122-1363 AG122-1332 CA123-1333 CAAG123-1332 CAAG122-1363 CAAG125-1333 CAAG125-1332
J&W DB-1301 Columns, Agilent Technologies
Chromatography Columns
A low- to mid-polarity column, equivalent to USP Phase G43.
  • (6%-Cyanopropyl-phenyl)-methylpolysiloxane
  • Low- to mid-polarity
  • Bonded, crosslinked, and solvent rinsable
  • Exact replacement of HP-1301
  • Equivalent to USP Phase G43
  • Tested with the tightest industry QC specifications for column bleed, sensitivity, and efficiency
  • Performance Summary shipped with each column
Agilent J&W DB-1301 (6% cyanopropyl-phenyl)-methylpolysiloxane column is a low- to mid-polarity, bonded, crosslinked, and solvent rinsable column, equivalent to USP phase G43.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty