Would you like to log in again?
J&W DB-VRX Columns, Agilent Technologies
Supplier: Agilent
J&W
122-1564 122-1564 124-1574 123-1564 122-1534 124-1534 123-1534 122-1564E 122-1564E 123-1564 124-1534 122-1534 123-1534
AG123-1564EA 1485.44 USD InStock
AG123-1564 AG123-1534 CAAG124-1534 CA122-1564 AG122-1564 AG122-1564E CA122-1534 AG122-1534 CAAG122-1564E CAAG123-1564 AG124-1534 CA123-1534 AG124-1574
J&W DB-VRX Columns, Agilent Technologies
Chromatography Columns
Features unique selectivity, engineered for optimum resolution of volatiles.
  • Unique selectivity engineered for optimum resolution of volatiles analysis
  • U.S. EPA Methods 502.2, 524.2 and 8260
  • 0.45 mm int.Ø columns provide more plates per meter compared to 0.53 mm id columns for the fewest co-elutions for GC method
  • Doesn't require subambient cooling to resolve the six "gases"
  • Fast run time: <30 minutes for optimum sample throughput, <8 minutes with 0.18 mm id
  • Low polarity
  • Excellent peak shape
  • Bonded, crosslinked, and solvent rinsable
  • Tested with the tightest industry QC specifications for column bleed, sensitivity, and efficiency
  • Performance Summary shipped with each column
Agilent J&W DB-VRX features unique selectivity, engineered for optimum resolution of volatiles. These low-polarity columns deliver excellent peak shapes and are fully compliant with U.S. EPA methods 502.2, 524.2, and 8260. They are bonded and crosslinked, and require no subambient cooling to resolve the six 'gases'. DB-VRX features an operating temperature range of –20 to 260 °C.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty