Would you like to log in again?
J&W DB-Dioxin Columns, Agilent Technologies
Supplier: Agilent Technologies
J&W
122-2462 122-2462
AG122-2462EA 1248 USD InStock
AG122-2462 CAAG122-2462
J&W DB-Dioxin Columns, Agilent Technologies
Chromatography Columns
Specifically engineered for the analysis of polychlorinated dibenzodioxins (PCDD) and dibenzofurans (PCDF).
  • Specifically engineered for the analysis of polychlorinated dibenzodioxins (PCDDs) and dibenzofurans (PCDFs)
  • Resolves 2,3,7,8-TCDD and 2,3,7,8-TCDF from all other isomers in one run
  • Low bleed, bonded, crosslinked, and solvent rinsable
  • Tested with the tightest industry QC specifications for column bleed, sensitivity, and efficiency
  • Performance Summary shipped with each column
Agilent J&W DB-Dioxin columns are specifically engineered for the analysis of polychlorinated dibenzodioxins (PCDD) and dibenzofurans (PCDF). The column resolves 2,3,7,8-TCDD and 2,3,7,8-TCDF from all other isomers in one run. A 100-psi regulator is required to reach optimum carrier gas velocity.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty