Would you like to log in again?
DB-5ms Ultra Inert Columns, Agilent Technologies
Supplier: Agilent
J&W
122-55H2UI 122-55H2UI 122-5563UI 122-5536UI 121-5523UI 122-5512UILTM 122-5522UI 165-6626 123-5536UI 122-5552UI 122-5532UI-INT 122-5512UI 122-5532UIE 122-5562UI 121-5522UI 122-5562UI-INT 122-5532UIE 122-5532UI 121-5512UI 122-5512UILTM 123-5532UI 122-5532UI-INT 123-5533UI 122-5533UI 123-5533UI 122-5512UI G3900-63005 121-5522UI 121-5522UI-INT 122-5532UI 121-5523UI 122-5563UI G3900-63005 123-5563UI 122-5562UI-INT 122-5536UI 122-5512UI-INT 121-5512UI 123-5563UI 121-5522UI-INT 122-5513UI 122-5552UI 165-6626 122-5562UI 122-5513UI 122-5522UI 123-5536UI 123-5532UI 122-5533UI
AG121-5512UIEA 435.44 USD InStock
AG121-5512UI AG1225512UILTM AG122-55H2UI AG123-5533UI CAAG122-55H2UI AG122-5512UI AG122-5533UI CA121-5522UI CA122-5562UI AG122-5552UI CAAG122-5533UI CAAG122-5552UI CAAG121-5512UI CAAG1215522UNT CAAG123-5533UI AG121-5522UI AG122-5563UI CAAG1225512UIL CA122-5532UI CA122-5536UI AG1215522UI CAAG121-5523UI CAAG123-5563UI CAAG122-5522UI AGG3900-63005 AG122-5536UI AG122-5513UI AG123-5536UI CAAG1225532UIE AG1225532UIINT AG123-5532UI AG122-5532UIE AG122-5532UI AG1225512UI AG1225562UIINT CAAG165-6626 CAAG123-5536UI CAAG122-5513UI CAAG1225562UNT CAAG123-5532UI AG121-5523UI AG165-6626 AG123-5563UI AG122-5522UI CAAG1225532UNT CA122-5512UI CAAGG390063005 AG122-5562UI CAAG122-5563UI
DB-5ms Ultra Inert Columns, Agilent Technologies
Chromatography Columns
Deliver consistent inertness, exceptionally low column bleed, great peak shapes, and effective performance for challenging active analytes.
  • Individually tested with Über One test probe mixture
  • Consistent column inertness performance
  • Exceptionally low column bleed
  • Great peak shapes for challenging active analytes such as pesticides and drugs of abuse
  • Excellent signal-to-noise ratios
  • Minimum compound adsorption or degradation
  • Support of 0.18 mm id column configuration for higher sample throughput
  • Tested with the tightest industry QC specifications for column bleed, sensitivity, and efficiency
  • Performance Summary shipped with each column
Agilent J&W DB-5ms Ultra Inert columns deliver industry-leading analytical performance in a range of chromatographic applications. DB-5ms Ultra Inert is low bleed and features excellent signal-to-noise ratio and great peak shapes, enabling high performance for challenging active analytes such as pesticides. DB-5ms Ultra Inert is tested with a demanding test probe mixture to ensure consistent column inertness.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty