Would you like to log in again?
J&W Intuvo Premium Polysiloxane Columns, Agilent Technologies
Supplier: Agilent
J&W
G3909-63006 G3909-63005 123-57J1-INT 122-5711-INT 122-1111-INT CP7483-INT 122-1111-INT 122-5731-INT G3909-63005 123-57J1-INT CP7483-INT 122-5731-INT CP7596-INT 122-5711-INT G3909-63006 CP7596-INT 123-1111-INT 123-1111-INT
AG122-1111-INTEA 666.03 USD InStock
AG122-1111-INT CAAGCP7483-INT CAAG1225731INT CAAG12357J1INT AG122-5711-INT CAAG1225711INT AG123-1111-INT AGG3909-63005 AGCP7483-INT AGCP7596-INT AG122-5731-INT AG123-57J1-INT AGG3909-63006 CAAG1231111INT CAAGG390963005 CAAG1221111INT CAAGG390963006 CAAGCP7596-INT
J&W Intuvo Premium Polysiloxane Columns, Agilent Technologies
Chromatography Columns
The most common stationary phases – stable, robust, and versatile; for use with the Intuvo 9000 GC System.
  • Wide selection of stationary phase polarities (low, mid, and high polarity)
  • Strict quality control specifications and testing ensure long-term column-to-column performance and accurate separations
  • Optimized GC flow path with Agilent Ultra Inert Liners, low-bleed septa, certified vials, and gas clean filters
  • For use with the Agilent Intuvo 9000 System
Agilent J&W Premium Polysiloxane Intuvo GC columns (compatible only with the Intuvo 9000 GC System) are most common stationary phases. Polysiloxanes are stable, robust, and versatile, and are available in the greatest variety. Standard polysiloxanes are characterized by the repeating siloxane backbone. Each silicon atom contains two functional groups; the type and percent level of substitution of the groups distinguish each stationary phase and its properties.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty