Would you like to log in again?
J&W DB-WAX Columns, Agilent Technologies
Supplier: Agilent
J&W
124-7032 127-7023E 128-7032 127-7023 123-7012 125-7062E 125-7012 123-7033 125-7032E 122-7032 127-7013 122-7033 123-7063E 127-7012 122-7032LTM 122-7012E 122-7013 128-7022 123-7032 123-7013 127-7023E 125-7037 125-7062E 125-7012E 127-7012 123-7063 122-7063 122-7062 122-7033E 122-7031 123-7033 122-7062E 128-7022 124-7032 122-7033 127-7013 123-7063 122-7032E 122-7033E 122-7062 123-7012 123-7032E 123-7033E 122-7032 123-7062 125-7017 125-7062 122-7061 122-7012 122-7061 125-7032E 125-7017 123-7063E 125-7012E 125-7031 125-7032 125-7037 122-7032E 122-7062E 123-7032 127-7022 125-7031 122-7012E 122-7013 123-7033E 125-7062 122-7031 122-7063 127-7023 123-7013 122-7012 122-7032LTM 125-7012 127-7022 128-7032 123-7062 128-7052 128-7052 123-7032E 125-7032
AG122-7062EA 1293.98 USD InStock
AG122-7062 AG128-7022 AG122-7061 CA125-7062 CAAG1227032LTM AG122-7033E AG127-7023E AG122-7063 CA123-7013 AG123-7032 CAAG125-7012E AG123-7033 AG127-7022 CA123-7012 CAAG127-7023E AG127-7023 AG125-7012E AG125-7032E AG128-7032 AG125-7031 AG125-7032 CA122-7012 AG123-7063E AG122-7032E CA123-7033E AG125-7037 CA123-7062 AG122-7012E AG122-7032LTM CA123-7063 AG122-7033 AG122-7032 AG122-7031 CAAG122-7031 CAAG125-7031 CAAG128-7032 AG127-7012 CAAG122-7062E AG127-7013 CAAG127-7023 CAAG127-7022 CAAG125-7037 CA125-7012 CAAG123-7032E CA122-7033E AG125-7062 AG125-7062E AG123-7032E CAAG124-7032 CAAG125-7062E AG123-7013 CAAG122-7063 CAAG122-7061 CA123-7032 CA123-7033 CA127-7012 CAAG128-7022 AG123-7012 CAAG127-7013 CA122-7062 CA122-7032E AG123-7033E CA122-7032 CAAG122-7012E CA122-7033 AG128-7052 AG125-7017 AG122-7013 AG122-7012 AG122-7062E AG125-7012 CA76514-554 CA125-7032E CAAG128-7052 AG124-7032 CAAG125-7017 AG123-7062 AG123-7063 CAAG122-7013 CAAG123-7063E
J&W DB-WAX Columns, Agilent Technologies
Chromatography Columns
A high-polarity polyethylene glycol (PEG) column that is ideal for food, fragrance, and flavor applications.
  • Polyethylene glycol (PEG) stationary phase
  • Close equivalent to USP Phase G16
  • High polarity
  • Temperature limit of 20 °C is lowest of any bonded PEG phase
  • Improves resolution of low boiling point analytes
  • High column-to-column reproducibility
  • Bonded, crosslinked, and solvent rinsable
  • Exact replacement of HP-WAX
  • Tested with the tightest industry QC specifications for column bleed, sensitivity, and efficiency
  • Performance Summary shipped with each column
Agilent J&W DB-WAX is ideal for food, fragrance, and flavor applications. The column's lower temperature limit of 20 °C improves resolution of analytes with low boiling points.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty