Would you like to log in again?
J&W DB-HeavyWax Columns, Agilent Technologies
Supplier: Agilent
J&W
122-7162 123-7133 123-7132-INT 123-7112 122-7163 122-7157 123-7162 122-7132-INT 127-7112 122-7162-INT 122-7132 121-7122 123-7112 123-7133-INT 123-7133 122-7127 123-7132 122-7133 123-7162 127-7112 122-7127 122-7163 122-7157 121-7122 123-7163-INT 123-7132-INT 122-7133 123-7133-INT 123-7163-INT 123-7163 122-7162 122-7162-INT 122-7132-INT 122-7112 122-7132 123-7132 123-7163 122-7112
AG122-7162-INTEA 1785 USD InStock
AG122-7162-INT AG123-7133-INT CAAG123-7162 CAAG1227162INT CAAG123-7163 AG123-7163-INT AG122-7163 AG122-7162 AG123-7112 CAAG122-7163 CAAG122-7162 CAAG1237132INT CAAG1227132INT AG123-7132 AG123-7132-INT AG123-7133 CAAG122-7127 CAAG127-7112 AG121-7122 CAAG123-7132 CAAG123-7133 CAAG123-7112 AG122-7157 CAAG121-7122 AG122-7112 AG122-7133 AG122-7132 CAAG1237163INT AG123-7163 AG122-7127 AG127-7112 CAAG122-7157 AG123-7162 CAAG1237133INT AG122-7132-INT CAAG122-7112 CAAG122-7133 CAAG122-7132
J&W DB-HeavyWax Columns, Agilent Technologies
Chromatography Columns
A high-temperature WAX column that can operate at temperatures of 280 °C isothermal and 290 °C programmed.
  • Polyethylene glycol selectivity
  • Maximum operating temperatures of 280 °C isothermal and 290 °C programmed
  • Fast analysis, with shorter run times due to extended temperature limit
  • Retention time stability and increased column lifetime, even when used at maximum operating temperature
  • Ability to bake out the column at the end of the analysis to reduce sample matrix effect and limit carryover and the risk of ghost peaks
  • Extended list of analytes to higher molecular weight compounds
  • Broader application range for multidimensional GC applications, as the oven temperature can be increased compared to standard WAX column
The Agilent J&W DB-HeavyWAX Polyethylene Glycol (PEG) is a high-temperature WAX column that can operate at higher maximum temperatures (280 °C isothermal, 290 °C programmed) than standard WAX columns. This allows for shorter run times, providing a lower cost per test. The column provides retention time stability and increased column lifetime, even when used at maximum operating temperature. The ability to bake out the column at the end of the analysis reduces sample matrix effects, limiting carryover and the risk of ghost peaks. The column also exhibits very low bleed, which results in better signal-to-noise ratios and lower detection limits (LOD), especially for late-eluting compounds.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty