Would you like to log in again?
J&W DB-WAXetr Columns, Agilent Technologies
Supplier: Agilent
J&W
122-7333 125-7314 123-7312 123-7363E 123-7333 125-7332 123-7354 125-7362 125-7334 123-7332 122-7332E 123-7364 123-7312 123-7332 122-7362E 123-7362 122-7363 123-7363 123-7354 125-7332E 123-7334 125-7312 122-7333E 122-7362E 122-7362 122-7332 125-7334E 125-7334 125-7334E 125-7362 125-7332 125-7332E 122-7332E 122-7333E 122-7332-INT 123-7333 125-7312 125-7314 125-7333 123-7363E 122-7363 122-7362 128-7323 122-7332 123-7363 122-7333 123-7364 123-7362 128-7323 122-7332-INT 125-7333 123-7334
AG125-7362EA 1418.52 USD InStock
CAAG122-7332E AG125-7362 CA122-7362 CAAG123-7363 CAAG123-7364 CAAG123-7362 AG122-7332E CA123-7333 AG123-7354 AG123-7312 CA123-7334 CAAG128-7323 CAAG122-7363 CAAG122-7333E CA122-7332 CA122-7333 CAAG123-7332 AG125-7314 AG125-7312 AG122-7332-INT AG123-7362 AG123-7363 AG123-7364 CAAG125-7334E CAAG125-7314 CA125-7334 CA125-7332 AG125-7332E CAAG125-7312 AG123-7363E CAAG1227332INT CAAG122-7362E AG122-7363 AG122-7362 AG128-7323 AG122-7362E AG123-7332 AG123-7333 AG123-7334 CAAG125-7362 CAAG123-7363E CAAG123-7312 AG122-7333E CAAG123-7354 AG122-7333 AG125-7334 AG122-7332 AG125-7332 AG125-7333 CAAG125-7332E AG125-7334E CAAG125-7333
J&W DB-WAXetr Columns, Agilent Technologies
Chromatography Columns
A high-polarity PEG column with an extended temperature range for alcohols, solvents, benzenes, styrenes, and organic acids.
  • Polyethylene glycol (PEG)
  • High polarity
  • Extended temperature range
  • Excellent column-to-column repeatability
  • Bonded, crosslinked, and solvent rinsable
  • Equivalent to USP phase G16
  • Tested against the strictest industry QC specifications for column bleed, sensitivity, and efficiency
  • Performance Summary shipped with each column
  • Similar phases: SUPELCOWAX 10, SUPEROX II, CB-WAX, Stabilwax, BP-20, 007-CW, Carbowax, Rtx-WAX, ZB-WAX, ZB-WAX plus
Agilent J&W DB-WAXetr is an extended temperature range high-polarity polyethylene glycol (PEG) column for the analysis of alcohols, solvents, benzenes, styrenes, organic acids, and more. It has excellent column-to-column repeatability and is closely equivalent to USP phase G16.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty