Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Vac Elut SPS 24 Manifold, Agilent Technologies
Supplier: Agilent Technologies
12234004 12234003 12234041 12234022 12234004 12234041 12234022 12234003
AG12234004EA 3827 USD InStock
AG12234004 CAAG12234041 CAAG12234022 AG12234041 AG12234003 CAAG12234004 AG12234022 CAAG12234003
Vac Elut SPS 24 Manifold, Agilent Technologies
Manifolds
Vac Elut 24 SPS manifold processes up to 24 SPE cartridges at the same time.
  • Closed operation prevents cross contamination
  • Stainless steel tips deliver maximum extract purity
  • Range of rack sizes covers most tube configurations
The Vac Elut SPS 24 manifold allows you to process up to 24 SPE cartridges simultaneously. Like all Vac Elut manifolds, the SPS 24 is made from durable, solvent-resistant materials and engineered to last. The glass sides allow easy viewing of the entire sample collection process.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty