Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
VacElut Cartridge Manifolds, Agilent Technologies
Supplier: Agilent Technologies
12234103 12234100 5982-9110 5982-9120 12234101 5982-9110 12234103 12234105 12234102 12234100 12234105 12234104 12234101 12234102 12234104
101478-540EA 1964.62 USD InStock
101478-540 AG5982-9110 AG12234105 AG12234104 AG12234103 CAAG5982-9110 AG12234102 AG12234101 AG12234100 CAAG12234105 CAAG12234100 CAAG12234103 CAAG12234104 CAAG12234101 CAAG12234102
VacElut Cartridge Manifolds, Agilent Technologies
Manifolds
VacElut cartridge manifolds enable simultaneous use of multiple Bond Elut cartridges.
  • Precise vacuum settings
  • Increased productivity/sample throughput
  • Eliminate cross contamination with disposable needles
  • Rugged, reliable construction
  • Transparent glass base allows observation of the whole collection operation
  • Simple vacuum adjustment
The corrosion-resistant VacElut cartridge manifolds allow simultaneous processing of multiple Bond Elut cartridges. The clear glass base allows observation of the entire sample collection process. Its compact, linear design requires little bench space.
These manifolds are also able to accommodate larger 16×150 mm test tubes. These collection vessels can be used in combinatorial chemistry applications using large boiling tubes for collection of purified synthesis mixtures. They can be used for any SPE extraction where an elution volume greater than 10 ml is required.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty