Would you like to log in again?
Bond Elut C2, Agilent Technologies
Supplier: Agilent Technologies
Bond Elut™
12256003 12256003
AG12256003PK 169 USD InStock
CAAG12256003 AG12256003
Bond Elut C2, Agilent Technologies
Chromatography Columns
Bond Elut C2 is mildly nonpolar, used where analytes are retained too strongly on C8 or C18 phases.
  • Low carbon load sorbent
  • Can be used alongside CN and C8 phases
  • Popular for drug extraction from plasma
The low carbon load of Bond Elut C2 creates a fairly nonpolar sorbent. C2 is often used during the process of method development if analytes are retained too strongly on a C8 or C18 phase. The polarity of C2 is slightly lower than a cyano phase for polar interactions.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty