Would you like to log in again?
Bond Elut PPL, Agilent Technologies
Supplier: Agilent Technologies
Bond Elut™
12105003 12105003 12256087 12256087
AG12105003PK 362 USD InStock
CAAG12256087 AG12105003 AG12256087 CAAG12105003
Bond Elut PPL, Agilent Technologies
Chromatography Columns
Bond Elut PPL is a modified styrene-divinylbenzene polymer that retains polar analytes.
  • Modified styrene-divinylbenzene polymer
  • Large particle size enables fast extraction speeds
  • High surface area and capacity for polar analytes
Bond Elut PPL (Priority PolLutant) is a styrene-divinylbenzene (SDVB) polymer modified with a proprietary nonpolar surface. PPL will retain even the most polar classes of analytes. The large particle size eases flow for viscous or particulate-rich water samples, while the large surface area and strong hydrophobicity ensure reproducible extractions with high recoveries.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty