Would you like to log in again?
Bond Elut AccuCAT, Agilent Technologies
Supplier: Agilent Technologies
Bond Elut™
12282002 12282005 12282001 12282004 12282001 12282005 12282004 12282002
AG12282005PK 312 USD InStock
AG12282005 CAAG12282001 CAAG12282002 AG12282001 CAAG12282004 AG12282002 CAAG12282005 AG12282004
Bond Elut AccuCAT, Agilent Technologies
Chromatography Columns
Bond Elut AccuCAT contains SAX and SCX sorbents to extract acidic, basic, and neutral analytes.
  • SCX and SAX functionalities offer broad analyte extraction potential
  • Ultraclean, mixed sorbent bed delivers for reproducible extractions
  • Compatible with many biological fluids for easy method transfer
Bond Elut AccuCAT cartridges are mixed-mode SPE packed bed cartridges consisting of a strong cation exchange (SCX) and a strong anion exchange (SAX) sorbent. AccuCAT is effective for the extraction of acidic, basic, and neutral analytes from biological samples, particularly for catecholamine extraction from biofluids.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty