Would you like to log in again?
J&W DB-1ms Ultra Inert Columns, Agilent Technologies
Supplier: Agilent
J&W
122-0162UI 122-0162UI 123-0132UI-INT 123-0132UI 123-0112UI 123-0132UI 122-0112UI 123-0132UI-INT 122-0112UI 123-0112UI 122-0132UI-INT 122-0132UI-INT 122-0132UI 122-0132UI
AG122-0112UIEA 516.76 USD InStock
CAAG1220132UNT AG122-0112UI AG123-0132UI AG123-0112UI CA122-0132UI AG1230132UIINT AG122-0132UI AG122-0162UI AG1220132UI CAAG122-0112UI CAAG122-0162UI CAAG1230132UNT CAAG123-0112UI CAAG123-0132UI
J&W DB-1ms Ultra Inert Columns, Agilent Technologies
Chromatography Columns
Provides consistent column inertness performance to ensure sensitivity and integrity in trace analysis.
  • Individually tested with Ultra Inert 1ms test probe mixture
  • Consistent column inertness performance and exceptionally low bleed
  • High-quality peak shapes for challenging active analytes
  • Excellent signal-to-noise ratios
  • Minimum compound adsorption or degradation
  • Supports 0.18 mm int.Ø column configuration for higher sample throughput
  • Performance Summary shipped with each column
Agilent J&W DB-1ms Ultra Inert delivers consistent column inertness performance to ensure analytical sensitivity and integrity. With excellent peak shapes, exceptionally low bleed, and high signal-to-noise ratios, this column performs effectively with even the most challenging active analytes. Each column is individually tested with Ultra Inert 1ms test probe mixture.
Tested against the strictest industry QC specifications for column bleed, sensitivity and efficiency.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty