Would you like to log in again?
J&W DB-1ht Columns, Agilent Technologies
Supplier: Agilent
123-1131E G3903-61006 123-1131 123-1111LTM 122-1111 122-1131 123-1111 125-1131 122-1131 122-1111 123-1111LTM 123-1131 125-1131 123-1102LTM 123-1102LTM G3903-61006 123-1111 123-1131E
AG123-1111EA 558.96 USD InStock
AG123-1111 AG123-1102LTM AG125-1131 CAAG1231111LTM CAAGG390361006 CAAG123-1131E AG123-1111LTM AG123-1131E AG122-1131 CA122-1131 CA123-1131 AG123-1131 CA122-1111 AG122-1111 CAAG125-1131 CAAG1231102LTM CA123-1111 AGG3903-61006
J&W DB-1ht Columns, Agilent Technologies
Chromatography Columns
100% PDMS specially processed for temperature limits of 400 °C, with excellent peak shape and faster elution times for high boilers.
  • 100% Dimethylpolysiloxane
  • Nonpolar
  • Specially processed for extended temperature limit of 400 °C
  • High temperature, polyimide-coated, fused silica tubing
  • Excellent peak shape and faster elution times for high boilers
  • Bonded, crosslinked, and solvent rinsable
  • Tested with the tightest industry QC specifications for column bleed, sensitivity, and efficiency
  • Performance Summary shipped with each column
Agilent J&W DB-1ht is a nonpolar, high-temperature 100% dimethlypolysiloxane column manufactured to the highest levels of quality and stability to deliver excellent performance in a range of GC/MS applications.It is specially processed for operation up to 400 °C, with a specially engineered high-temperature polyimide-coated fused silica tubing.
This bonded, crosslinked, solvent-rinsable column delivers excellent peak shape and faster elution times for high-boiling compounds.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty