Would you like to log in again?
J&W DB-35 Columns, Agilent Technologies
Supplier: Agilent
J&W
122-1932 125-1937 123-1968 123-1932 123-1968 123-1933 123-1933E 125-1932 123-1932 122-1932 125-1932 125-1937 123-1933E 123-1933
AG123-1968EA 1300.15 USD InStock
CAAG123-1933E AG123-1968 AG123-1932 CA123-1932 CAAG122-1932 AG123-1933 AG123-1933E CAAG125-1932 CAAG125-1937 CAAG123-1968 AG125-1932 AG125-1937 AG122-1932 CAAG123-1933
J&W DB-35 Columns, Agilent Technologies
Chromatography Columns
A mid-polarity column excellent for environmental analyses.
  • (35%-Phenyl)-methylpolysiloxane
  • Mid polarity - slightly more polar than HP-35
  • Inert to active solutes
  • Ideal for confirmational analyses
  • Bonded, crosslinked, and solvent rinsable
  • Equivalent to USP Phase G42
  • Tested with the tightest industry QC specifications for column bleed, sensitivity, and efficiency
  • Performance Summary shipped with each column
Agilent J&W DB-35 columns are ideal for conformational analysis. The DB-35 is a (35%-phenyl)-methylpolysiloxane, mid-polarity column. Bonded and crosslinked, it is inert to active solutes. DB-35 is a close equivalent to USP phase G42.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty