Would you like to log in again?
J&W DB-23 Columns, Agilent Technologies
Supplier: Agilent
J&W
123-2332 125-2312 122-2361-INT 122-2361 122-2361-INT 122-2312 125-2312 122-2332E 125-2332 123-2362 122-2362E 122-2362 122-2362 122-2332E 123-2332 122-2331 125-2332 122-2332-INT 123-2362 122-2332 122-2312 122-2332 122-2331 122-2362E 122-2361 122-2332-INT
AG123-2332EA 810.15 USD InStock
CAAG123-2332 CAAG122-2332E CAAG1222332INT CAAG1222361INT AG123-2332 AG125-2332 CAAG122-2362E AG122-2332-INT CA122-2312 AG123-2362 CA122-2332 CAAG122-2331 CAAG125-2312 CAAG125-2332 CAAG123-2362 AG122-2362 AG122-2361 AG122-2312 AG122-2331 AG125-2312 AG122-2332 AG122-2361-INT AG122-2362E CA122-2362 CA122-2361 AG122-2332E
J&W DB-23 Columns, Agilent Technologies
Chromatography Columns
A high-polarity, bonded, and solvent-rinsable stationary phase for the separation of fatty acid methyl esters (FAMEs).
  • (50%-Cyanopropyl)-methylpolysiloxane
  • High polarity, replaces HP-23
  • Designed for separation of fatty acid methyl esters (FAMEs)
  • Excellent resolution for cis- and trans-FAME isomers
  • Close equivalent to USP phase G5
  • Bonded, crosslinked, and solvent rinsable
  • Tested with the tightest industry QC specifications for column bleed, sensitivity, and efficiency
  • Performance Summary shipped with each column
Agilent J&W DB-23 is specifically designed for the high-quality separation of fatty acid methyl esters (FAMEs). It is bonded, crosslinked, and solvent rinsable, and is a close equivalent to USP phase G5. This column has excellent resolution and a wide operating temperature range, and is well suited for procedures using cis- and trans- FAME isomers.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty