Would you like to log in again?
DB-FFAP Columns, Agilent Technologies
Supplier: Agilent
J&W
123-3233 125-3231 125-3212 123-3212 127-32H2 127-3212 125-3231 122-3212 122-3263 123-3233E 125-3233 122-3232E 122-3233 125-3217 123-3263 123-3232E 125-3262 125-3237 123-3263 123-3212 123-3253 125-3237 122-3232E 122-3212 122-3262 122-3233 123-3233E 123-3232E 123-3264 125-3217 123-3223 123-3233 123-3253 123-3264 125-3232 127-32H2 122-3262 123-3234 125-3232E 123-3262 125-3232 123-3232 122-3263 123-3232 125-3233 122-3232 125-3212 127-3212 122-3232 124-3232 124-3232 123-3234 123-3262 123-3223 125-3262
AG124-3232EA 901.76 USD InStock
AG124-3232 AG127-3212 CAAG125-3217 AG122-3263 AG122-3262 AG123-3223 AG122-3212 AG123-3232 AG125-3237 AG125-3232E CAAG125-3262 CAAG123-3212 CAAG123-3253 AG125-3231 AG125-3232 AG125-3233 AG123-3233 AG123-3234 CA123-3232E AG122-3232E CAAG123-3233E CAAG125-3233 CAAG125-3231 CAAG123-3262 CAAG123-3263 CAAG125-3237 CAAG122-3212 CAAG123-3223 CAAG123-3264 AG125-3262 AG125-3217 CAAG124-3232 AG123-3253 CA123-3232 AG122-3232 CAAG127-3212 AG122-3233 AG123-3232E CAAG122-3263 CA123-3234 CAAG122-3262 CAAG123-3233 CAAG127-32H2 AG125-3212 AG123-3212 AG123-3233E AG123-3264 CAAG122-3232E CA122-3232 AG123-3262 AG123-3263 CA122-3233 AG127-32H2 CA125-3232 CAAG125-3212
DB-FFAP Columns, Agilent Technologies
Chromatography Columns
DB-FFAP is designed for the analysis of volatile fatty acids and phenols.
  • Nitroterephthalic acid modified polyethylene glycol
  • High polarity
  • Designed for the analysis of volatile fatty acids and phenols
  • Bonded and cross-linked
  • solvent rinsable
  • Close equivalent to USP Phase G35
  • Replaces OV-351
  • Agilent J&W GC columns are tested against the strictest industry QC specifications for column bleed, sensitivity, and efficiency to give you utmost confidence in qualitative and quantitative results
  • For QC test conditions, including the test probe mixture and the resulting chromatogram, refer to the Agilent J&W GC Column Performance Summary shipped with each GC column
  • Note: We do not recommend the use of water or methanol to rinse DB-FFAP GC columns
  • Similar Phases: SPB-5, Rtx-5, BP-5, OV-5, 007-2(MPS-5), SE-52, SE-54, XTI-5, PTE-5, ZB-5, AT-5, MDN-5, ZB-5
Agilent J&W DB-FFAP is a nitroterephthalic-acid-modified polyethylene glycol (PEG) column of high polarity for the analysis of volatile fatty acids and phenols. It is a close equivalent to USP phase G35 and replaces OV-351.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty