Would you like to log in again?
J&W Ultra Inert DB-35ms Columns, Agilent Technologies
Supplier: Agilent
122-3832UI 121-3822UI 122-3812UI-INT 123-3832UI-INT 122-3812UI-INT 123-3832UI-INT 122-3812UI 122-3832UI 121-3822UI 122-3812UI 123-3832UI 122-3832UI-INT 123-3832UI 122-3832UI-INT
AG1223832UIINTEA 1035.6 USD InStock
AG1223832UIINT AG123-3832UI AG122-3832UI AG121-3822UI AG122-3812UI AG1223812UI CAAG121-3822UI CA122-3832UI AG1233832UIINT CA122-3812UI CAAG1223832UNT CAAG123-3832UI CAAG1233832UNT CAAG1223812UNT
J&W Ultra Inert DB-35ms Columns, Agilent Technologies
Chromatography Columns
Improved inertness and exceptionally low bleed, with great peak shapes for challenging active analytes such as pesticides and drugs of abuse.
  • Individually tested with the Ultra Inert DB-35ms test probe mixture
  • Consistent column inertness performance
  • Exceptionally low column bleed
  • Great peak shapes for challenging active analytes, with excellent signal-to-noise ratios
  • Minimum compound adsorption or degradation
  • Supports 0.18 mm int.Ø column configuration for higher sample throughput
Agilent J&W DB-35ms Ultra Inert is a mid-polarity column for applications where different selectivity is needed for enhanced separations, including pesticide analysis and drugs of abuse testing. This column quantifies components at a lower level, with broader linearity and optimized separation, ultimately increasing productivity.
Tested with the tightest industry QC specifications for column bleed, sensitivity, and efficiency.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty