Would you like to log in again?
DB-17ms Columns, Agilent Technologies
Supplier: Agilent
J&W
122-4732E 122-4732 123-4732 122-4712 122-4762 G3900-65001 G3900-65001 123-4732 122-4712 121-4722 123-4732-INT 122-4712LTM 123-4732-INT 122-4732 122-4711 122-4712LTM 121-4722 122-4762 123-4712 122-4731 122-4732E 122-4731 122-4711 123-4712
AG121-4722EA 696.9 USD InStock
CAAG122-4732E CAG3900-65001 CA121-4722 CAAG122-4711 AG121-4722 AG123-4732 AG123-4732-INT AG123-4712 CA122-4712 AG122-4712 CA122-4732 AG122-4732 AG122-4711 AG122-4731 CA122-4731 AG122-4762 CA123-4732 AGG3900-65001 CAAG123-4712 CAAG122-4762 CAAG1234732INT AG122-4732E AG122-4712LTM CAAG1224712LTM
DB-17ms Columns, Agilent Technologies
Chromatography Columns
Mid-polar GC/MS column with enhanced mass spectral integrity and excellent inertness for active compounds, particularly pesticides.
  • Virtually equivalent to (50%-phenyl)-methylpolysiloxane
  • Very low bleed and mid-polarity, ideal for GC/MS
  • Excellent inertness for active compounds
  • Enhanced mass spectral integrity
  • Bonded, crosslinked, and solvent rinsible
  • The best column for CLP pesticides
  • Tested with the tightest industry QC specifications for column bleed, sensitivity, and efficiency
  • Performance Summary shipped with each column
  • Similar phases: Rxi-17Sil MS, Rtx-50, 007-17, SP-2250, SPB-50, BPX-50, SPB-17, AT-50
Agilent J&W DB-17ms, a mid-polar column virtually equivalent to (50%-phenyl)-methylpolysiloxane, delivers excellent levels of inertness for the analysis of active compounds. The column is bonded, crosslinked, and solvent rinsible, and is excellent for the analysis of pesticides and other semi volatiles. Very low bleed performance makes it ideal for a range of GC/MS applications, for robust and accurate performance every time.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty