Would you like to log in again?
J&W Ultra Inert DB-BAC1 and DB-BAC2 Columns, Agilent Technologies
Supplier: Agilent
J&W
125-9334UI 123-9434UI 123-9334UI 123-9334UI 123-9434UI 125-9334UI
AG123-9434UIEA 851.32 USD InStock
AG123-9434UI AG123-9334UI AG125-9334UI CAAG125-9334UI CAAG123-9334UI CAAG123-9434UI
J&W Ultra Inert DB-BAC1 and DB-BAC2 Columns, Agilent Technologies
Chromatography Columns
Agilent DB-BAC1 Ultra Inert and DB-BAC2 Ultra Inert columns are able to provide separation of all important blood alcohol compounds.
  • Superior solution for accurate blood alcohol quantification
  • Optimized resolution and baseline separation of critical blood alcohol peaks
  • Excellent peak shape and accurate integration of low-concentration compounds
  • Accurate identification of challenging polar analytes, even at trace levels
  • Each column is tested with a uniquely designed QC test mixture to verify column performance
  • Rapid separation in just over 2 minutes using either t‑Butanol or n‑Propanol as an internal standard
  • Agilent Blood Alcohol Checkout Mix to verify the performance of the blood alcohol workflow
Agilent J&W Ultra Inert DB-BAC1 and DB-BAC2 columns are part of the Agilent J&W Ultra Inert GC column family capable of separating t-Butanol and n-Propanol from frequently encountered interferences in blood matrix. The unique polarity of the columns provides excellent selectivity when using this column set in GC/FID/FID analysis.
Agilent ethanol calibration standards for quantitative blood alcohol analysis, traceable to both NIST and European Reference materials (ERM). Similar phases: Rtx-BAC Plus 1, Rtx-BAC Plus 2, ZB-BAC-1, and ZB-BAC-2.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty