Would you like to log in again?
Biodiesel Columns, Agilent Technologies
Supplier: Agilent Technologies
123-BD34 123-BD01 123-BD11 160-BD65-5 123-BD11 160-BD65-5 1909BD-113 123-BD34 123-BD01 1909BD-113
AG123-BD11EA 559 USD InStock
CAAG123-BD11 CAAG160-BD65-5 AG123-BD11 CAAG123-BD34 CAAG123-BD01 AG123-BD34 AG123-BD01 AG1909BD-113 CAAG1909BD-113 AG160-BD65-5
Biodiesel Columns, Agilent Technologies
Chromatography Columns
Designed and application-optimized for the analysis of biodiesel to meet ASTM and EN testing standards.
  • Complete range of biodiesel GC columns specially designed according to EN 14105, ASTM D6584, EN 14103, EN 14110, and EN 14331 methods
  • Bonded and crosslinked
  • Tested with the tightest industry QC specifications for column bleed, sensitivity, and efficiency
  • Performance Summary shipped with each column
Agilent J&W Biodiesel is designed and application-optimized for the analysis of biodiesel to meet ASTM and EN testing standards, including ASTM D6584, EN 14013, EN 14105, EN 14106, and EN 14110. Engineered to meet the growing demand for analysis of biofuels as supplements or replacements for petroleum-based fuels, the column provides the highest level of performance for the determination of free and total glycerin, FAMEs, and trace methanol.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty