Would you like to log in again?
J&W Ultra Inert DB-WAX Columns, Agilent Technologies
Supplier: Agilent
123-7032UI-INT 123-7062UI 125-7032UI 122-7033UI-INT 123-7063UI 122-7062UI 123-7062UI 125-7012UI 123-7032UI 128-7022UI 123-7012UI 125-7012UI 122-7032UI-INT 123-7033UI-INT 125-7032UI 125-7031UI 122-7063UI 122-7012UI 123-7063UI 122-7033UI 123-7033UI 122-7032UI 122-7062UI 125-7031UI 122-7062UI-INT 128-7022UI 125-7037UI 123-7033UI-INT 122-7032UI 122-7062UI-INT 122-7032UI-INT 122-7012UI 122-7033UI 123-7033UI 123-7032UI 125-7037UI 123-7032UI-INT 123-7012UI 122-7063UI
AG1237033UIINTEA 1067.5 USD InStock
CA122-7032UI CA122-7033UI CA123-7033UI AG1237033UIINT AG1227032UI AG125-7031UI AG125-7032UI AG1227033UI AG125-7012UI AG123-7032UI AG123-7033UI CAAG1227032UNT CAAG128-7022UI AG1227062UIINT AG125-7037UI CAAG1237032UNT AG1237032UIINT CAAG123-7063UI CAAG123-7062UI CAAG122-7062UI CAAG122-7063UI CAAG122-7012UI CAAG123-7012UI CAAG1227062UNT AG123-7063UI AG122-7062UI AG123-7062UI AG122-7063UI AG128-7022UI AG122-7012UI AG122-7032UI AG123-7012UI AG122-7033UI CAAG1237033UNT CAAG125-7012UI CAAG125-7031UI CAAG125-7032UI CAAG123-7032UI CAAG125-7037UI
J&W Ultra Inert DB-WAX Columns, Agilent Technologies
Chromatography Columns
Deliver the highest levels of PEG stationary phase inertness performance for a range of challenging active analytes.
  • Improved peakshape for challenging polar compounds with DB-WAX Ultra Inert
  • Reliable column-to-column inertness performance
  • Bonded polyethylene glycol (PEG) selectivity equivalent to DB-WAX
  • No method revalidation needed
  • High polarity
  • Equivalent to USP G16
  • Excellent performance for acidic compounds, including free fatty acids – no need for FFAP type column
  • Bonded and crosslinked phase is solvent rinseable and tolerates aqueous injections
DB-WAX Ultra Inert (UI) represents the ultimate performance of PEG stationary phase inertness, providing symmetric peak shape and improved response for challenging polar compounds. All DB-WAX UI columns are individually tested with an Ultra Inert Wax test probe mixture according to the strictest industry QC standards to ensure consistent column-to-column inertness performance for challenging active analytes such as free fatty acids, alcohols, diols, and aldehydes, with minimal compound adsorption or degradation. Since it offers the same selectivity as Agilent J&W DB-WAX, upgrading to Ultra Inert performance is easy, with minimal validation required – there's no need to recreate or modify existing compound libraries that are based on DB-WAX selectivity.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty