Would you like to log in again?
J&W Ultra Inert Intuvo Columns, Agilent Technologies
Supplier: Agilent
123-0334UI-INT 122-9732-INT 123-9334UI-INT 123-0334UI-INT G3909-63004 123-9334UI-INT G3909-63004 122-0334UI-INT 123-9434UI-INT 123-9434UI-INT 122-9732-INT 122-0334UI-INT
AGG3909-63004EA 1062.36 USD InStock
CAAG1229732INT CAAG1239334UNT AGG3909-63004 CAAG1230334UNT AG1239334UIINT CAAG1220334UNT AG1230334UI AG1239434UIINT AG122-9732-INT CAAGG390963004 AG1220334UI CAAG1239434UNT
J&W Ultra Inert Intuvo Columns, Agilent Technologies
Chromatography Columns
Provide excellent inertness, low-bleed performance, and detection limits with the Intuvo 9000 GC system.
  • Wide selection of stationary phase polarities are available (non, low, mid, and high polarity)
  • Strict quality control specifications and testing ensure long-term column-to-column performance and accurate separations
  • Optimized GC flow path with Agilent Ultra Inert Liners, low-bleed septa, certified vials, and gas clean filters
  • For use with the Agilent Intuvo 9000 GC System
As part of the Agilent J&W Ultra Inert GC column family, Ultra Inert Intuvo GC columns (compatible only with the Intuvo 9000 GC System) push industry standards for consistent column inertness and exceptionally low column bleed, resulting in lower detection limits and more accurate data for difficult analytes. Offered in a wide range of stationary phase polarities (non, low, mid, and high polarity), each Ultra Inert Intuvo GC column is tested with the industry’s most demanding test probe mixture, and a performance summary sheet is shipped with each column.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty