Would you like to log in again?
J&W DB-Select 624 UI <467> Columns, Agilent Technologies
Supplier: Agilent
J&W
125-0334UI 123-0364UI 122-0334UI 123-0364UI 122-0334UI 123-0334UI 123-0334UI 125-0334UI
AG123-0334UIEA 934.72 USD InStock
AG123-0334UI CA122-0334UI AG122-0334UI AG125-0334UI CAAG125-0334UI CAAG123-0364UI CA123-0334UI AG123-0364UI
J&W DB-Select 624 UI <467> Columns, Agilent Technologies
Chromatography Columns
Designed for residual solvent analysis of pharmaceutical compounds per US Pharmacopeia Method <467>.
  • Engineered to optimize pharmaceutical residual solvents analysis per USP Method <467>
  • Ultra inertness and low bleed
  • Resolution of USP regulated critical pairs, also separates benzene and 1,2-dichloroethane
  • Identical selectivity to the popular VF-624 ms – upgrade with no changes in method
  • UI testing ensures premium performance column to column
  • Tested against the strictest industry QC specifications for column bleed, sensitivity and efficiency
  • Performance Summary shipped with each column
Agilent J&W DB-Select 624 UI <467> is designed specifically for United States Pharmacopoeia Method <467>. Equivalent to USP phase G43, this stationary phase is engineered for the best sensitivity and resolution of the critical pairs specified in <467> for the analysis of residual solvents in active pharmaceutical ingredients.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty