Would you like to log in again?
J&W DB-1701 Columns, Agilent Technologies
Supplier: Agilent Technologies
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
CrossLab J&W
127-0722 123-0753 125-0732 127-0723 125-0731 123-0763 123-0733 123-0762 123-0763 122-0732 122-0762 123-0733E 123-0733-INT 122-0762 123-0733 123-0732 125-0733 125-0733 123-0733E 123-0753 127-0723 122-0733 125-0712 123-0712 123-0733-INT 127-0722 123-0731 125-0762 125-0712 125-0731 121-0722 123-0713 123-0762 123-0732E 122-0732 123-0732E 125-0762 122-0733 123-0712 123-0713 121-0722 123-0732 123-0731 125-0732
AG123-0712EA 455 USD InStock
CA123-0712 AG123-0712 AG123-0733 CA123-0733 CAAG125-0762 AG123-0713 AG123-0732 AG123-0753 AG123-0731 CAAG123-0732E CAAG123-0733E AG122-0762 AG125-0762 AG123-0733E CA125-0712 CA125-0732 CAAG1230733INT CAAG123-0762 CA123-0763 AG123-0733-INT CAAG125-0731 CAAG125-0733 CAAG121-0722 AG123-0763 CA122-0733 CA122-0732 AG123-0762 CAAG127-0722 CAAG127-0723 CA122-0762 CAAG123-0713 AG125-0712 AG125-0733 AG121-0722 AG125-0732 AG125-0731 AG123-0732E AG127-0722 AG127-0723 AG122-0732 CAAG123-0753 AG122-0733 CAAG123-0731 CA123-0732
J&W DB-1701 Columns, Agilent Technologies
Chromatography Columns
A low/mid polarity column ideal for the analysis of pesticides, herbicides, TMS sugars, and aroclors.
  • (14%-Cyanopropyl-phenyl)-methylpolysiloxane
  • Solvent rinsable with low/mid polarity
  • Bonded and crosslinked
  • Exact replacement of HP-1301 and HP-1701
  • Available with DuraGuard (the GC column with a built-in guard column)
  • Tested against the strictest industry QC specifications for column bleed, sensitivity, and efficiency
  • Performance Summary shipped with each column
  • Similar phases: SPB-1701, Rtx-1701, BP-10, OV-1701, 007-1701, ZB-1701
Agilent J&W DB-1701 is a low/mid-polarity (14% cyanopropyl-phenyl)-methylpolysiloxane column. It is ideal for the analysis of pesticides, herbicides, trimethylsilyl (TMS) derivatives of sugars and related substances, and aroclors. The column is bonded, crosslinked, and solvent rinsable and is available with DuraGuard (the GC column with a built-in guard column).
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty