Would you like to log in again?
J&W Ultra Inert DB-624 Columns, Agilent Technologies
Supplier: Agilent Technologies
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
J&W
122-1364UI 123-1334UI-INT 123-1364UI 122-1364UI-INT 125-1374UI 122-1334UI-INT 122-1334UI 122-1364UI-INT 127-1314UI 123-1334UI 125-1334UI 123-1364UI 125-1374UI 122-1334UI 123-1334UI-INT 121-1324UI 122-1334UI-INT 127-1314UI 123-1364UI-INT 121-1324UI 123-1334UI 123-1364UI-INT 125-1334UI 122-1364UI
AG125-1374UIEA 1840 USD InStock
CA122-1334UI CA121-1324UI AG125-1374UI CAAG1221364UNT CA123-1334UI AG1221334UIINT AG1231334UI CAAG1231364UNT AG127-1314UI AG1231364UIINT CAAG1231334UNT CAAG1221334UNT AG1221364UIINT CAAG123-1364UI CAAG125-1374UI AG121-1324UI AG122-1334UI CAAG127-1314UI AG123-1334UI CA125-1334UI AG125-1334UI CA122-1364UI AG123-1364UI AG122-1364UI
J&W Ultra Inert DB-624 Columns, Agilent Technologies
Chromatography Columns
Designed for trace analysis in environmental, pharmaceutical, and other volatile organic compound (VOC) applications.
  • Environmental volatile organic compounds (VOCs) methods
  • Excellent for US EPA Methods: 501.3, 502.2, 503.1, 524.2, 601, 602, 8010, 8015, 8020, 8240, and 8260
  • Industrial chemical analysis – solvents, petrochemicals, specialty chemicals
  • Food and beverage – alcohols, fusel oils
  • Pharmaceutical residual solvents per USP
  • Ultra inertness processing expands application range with excellent peak shape for low molecular weight acidic compounds
  • UI testing ensures premium performance column-to-column
  • Identical selectivity to the industry standard DB-624 – upgrade with no change in method required
  • Optimized by the inventors of DB-624
Agilent J&W DB-624 UI is optimal for the fast analysis of volatile organic compounds (VOCs) in pharmaceutical, environmental, food, and industrial chemical applications. The unique Ultra Inert deactivation process enhances peak shape, improving signal-to-noise levels and increasing sensitivity for qualitative and quantitative analysis.
Ultra inertness processing expands application range with excellent peak shape for low molecular weight acidic compounds. UI testing ensures premium performance column-to-column. Identical selectivity to the industry standard DB-624 – upgrade with no change in method required.
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty