Would you like to log in again?
J&W DB-624 Columns, Agilent Technologies
Supplier: Agilent
125-1374E 124-1374 124-1334 123-1364E 121-1324E 123-1364E 125-1314 125-13A4 123-1334-KEY 123-1364 123-1364-KEY 125-1334-KEY 121-1324E 123-1334 123-1364 122-1364E 122-1364E 128-1324E 128-1324 122-1334-KEY 125-1374 125-1364 124-1374 125-1364 125-1334E 123-1334-KEY 122-1334-KEY 122-1334E 128-1324 125-1364-KEY 125-1334 125-1374 125-1334-KEY 128-1324E 123-1364-KEY 123-1334E 123-1334 122-1364-KEY 124-1334 125-13B4 125-1334E 125-1334 123-1334E 125-13B4 122-1364 125-1374E 122-1334 122-1334E 125-1314 122-1364 125-13A4 122-1364-KEY 125-1364-KEY 125-1364E 122-1334 125-1364E
AG124-1374EA 1713.97 USD InStock
AG124-1374 CAAG125-1314 AG123-1364 CA122-1364E CAAG121-1324E AG128-1324E AG125-1334 CAAG125-13B4 CA76514-550 AG122-1334-KEY AG123-1334E CA125-1364 CAAG128-1324E CAAG125-1364E AG125-1334E CAAG1251364KEY AG125-1374E AG125-1374 AG123-1334 CA123-1334E AG122-1334E AG128-1324 AG121-1324E AG122-1364 AG123-1364-KEY CAAG1231364KEY AG123-1334-KEY CA123-1364 AG125-13B4 CA122-1334E AG125-1364 CAAG1231334KEY CAAG1221334KEY CAAG1251334KEY AG122-1334 AG123-1364E CA128-1324 CAAG124-1374 AG122-1364-KEY AG125-1314 CA122-1364 AG125-1364E AG125-13A4 CA123-1364E AG125-1334-KEY AG122-1364E AG125-1364-KEY CA76514-552 CAAG124-1334 CA122-1334 CAAG125-13A4 CA125-1334E CAAG1221364KEY CA125-1374E CA125-1374 AG124-1334
J&W DB-624 Columns, Agilent Technologies
Chromatography Columns
Recommended for EPA methods 502.2 and 8021, as well as for fast GC/MS analysis of volatiles.
  • Specifically designed for the analysis of volatile priority pollutants and residual solvents
  • Bonded, crosslinked, and solvent rinsable
  • Equivalent to USP Phase G43
  • Exact replacement of HP-624
  • Available with DuraGuard (GC column with a built-in guard column)
  • No cryogenics needed for U.S. EPA Method 502.2
  • Excellent for U.S. EPA Methods 501.3, 502.2, 503.1, 524.2, 601, 602, 8010, 8015, 8020, 8240, 8260 and USP 467
  • Excellent inertness for active compounds
  • Tested with the tightest industry QC specifications for column bleed, sensitivity, and efficiency
  • Performance Summary shipped with each column
Agilent J&W DB-624 is recommended for EPA methods 502.2 and 8021, as well as for fast GC/MS volatiles analysis. It is specifically designed for the analysis of volatile priority pollutants and residual solvents.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty