Would you like to log in again?
DB-17 Columns, Agilent Technologies
Supplier: Agilent Technologies
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
CrossLab J&W
127-1722 125-1704 125-1704 127-1722 123-1712 127-1712 125-1733 123-1711 123-1711 127-1712 123-1733 125-1731 122-1733 122-1732E 125-1712 123-1713 123-1762 122-1732E 122-1762 123-1733 125-1732E 125-1762 122-1713 122-1732LTM 125-1713 123-1732-INT 125-1717 123-1732 125-1762 122-1713 122-1712 125-1732 123-1713 122-1762 125-1712 123-1762 125-1732E 122-1732LTM 125-1731 125-1717 122-1732 122-1712 123-1712 125-1713 123-1732 125-1733 123-1732-INT 125-1732 122-1733 122-1731 122-1731 122-1732
AG123-1732-INTEA 945 USD InStock
CAAG122-1731 AG123-1732-INT CAAG122-1733 CAAG125-1713 CAAG125-1712 AG123-1762 CAAG125-1717 CAAG127-1722 AG125-1731 AG125-1733 AG125-1732 CAAG123-1733 CAAG125-1762 AG122-1712 AG122-1713 CAAG122-1762 CAAG125-1732E AG123-1732 AG123-1733 CAAG127-1712 AG122-1762 AG125-1762 CAAG122-1732E AG127-1712 CAAG125-1733 CAAG1221732LTM CAAG125-1731 CAAG122-1713 CA122-1712 AG122-1732E CAAG1231732INT AG122-1732 AG122-1731 AG125-1713 AG125-1712 AG125-1717 CAAG123-1711 CA123-1732 AG122-1733 AG127-1722 CAAG123-1713 CAAG123-1712 AG123-1712 AG123-1713 CAAG125-1704 AG125-1732E AG123-1711 CA125-1732 AG122-1732LTM CA122-1732 CAAG123-1762 AG125-1704
DB-17 Columns, Agilent Technologies
Chromatography Columns
A mid-polarity column that is excellent for confirmation analysis.
  • (50%-Phenyl)-methylpolysiloxane
  • Mid polarity – slightly more polar than HP-50+
  • Excellent for confirmation analysis
  • Bonded, crosslinked, and solvent rinsible
  • Equivalent to USP phase G3
  • Tested with the tightest industry QC specifications for column bleed, sensitivity, and efficiency
  • Performance Summary shipped with each column
  • Similar phases: Rtx-50, 007-17(MPS-50), SP-2250, SPB-50, ZB-50, AT-50
Agilent J&W DB-17 is a (50%-phenyl)-methylpolysiloxane column of mid polarity. Equivalent to USP phase G3, it is excellent for confirmation analysis.
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty