Would you like to log in again?
J&W DB-608 Columns, Agilent Technologies
Supplier: Agilent
125-1730 123-1730 125-6837 125-6837 125-1730 122-6832 123-1730 122-6832
AG125-1730EA 825.59 USD InStock
AG125-1730 AG125-6837 CA125-6837 AG122-6832 CA125-1730 CAAG122-6832 CAAG123-1730 AG123-1730
J&W DB-608 Columns, Agilent Technologies
Chromatography Columns
Specifically designed for the analysis of chlorinated pesticides and PCBs, suitable for use in EPA methods 608, 508, and 8080.
  • Specifically designed for the analysis of chlorinated pesticides and PCBs
  • US EPA Methods 608, 508, and 8080
  • Excellent inertness and recovery without pesticide breakdown
  • Bonded, crosslinked, and solvent rinsable
  • Tested with the tightest industry QC specifications for column bleed, sensitivity, and efficiency
  • Performance Summary shipped with each column
Agilent J&W DB-608 columns are precision-engineered to deliver industry-leading performance in the analysis of chlorinated pesticides and PCBs. Widely used for US EPA methods 608, 508, and 8080, they deliver excellent inertness and recovery, without resulting in pesticide breakdown. This column is bonded, crosslinked, and solvent rinsable, with operating temperature limits of 40 to 280/300 °C.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty