Would you like to log in again?
J&W DB-200 Columns, Agilent Technologies
Supplier: Agilent Technologies
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
122-2032LTM 123-2032 122-2033 122-2033 122-2032 123-2033 122-2032LTM 122-2032 123-2032 125-2032 123-2033 125-2032
AG123-2032EA 787 USD InStock
AG123-2032 AG122-2033 AG123-2033 CAAG123-2032 AG122-2032LTM CAAG123-2033 CAAG122-2032 AG125-2032 CAAG125-2032 AG122-2032 CAAG122-2033 CAAG1222032LTM
J&W DB-200 Columns, Agilent Technologies
Chromatography Columns
A mid-polarity column ideal for difficult-to-separate positional isomers.
  • Temperature limits of 300 °C isothermal and 320 °C programmed
  • Mid polarity, more polar than DB-1701 or DB-17
  • Low ECD bleed and unique selectivity
  • Close equivalent to USP phase G6
  • Tested with the tightest industry QC specifications for column bleed, sensitivity, and efficiency
  • Performance Summary shipped with each column
Agilent J&W DB-200 is a (35% trifluoropropyl)-methylpolysiloxane column. This mid-polarity column is ideal for difficult-to-separate positional isomers. It has unique interactions with compounds containing nitro, halogen, and carbonyl groups.
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty