Would you like to log in again?
J&W DB-225 Columns, Agilent Technologies
Supplier: Agilent
J&W
122-2231 122-2232-INT 125-2212 125-2237 122-2232 123-2232 122-2232 123-2232 125-2237 125-2212 123-2232-INT 122-2212 122-2212 125-2232 125-2232 122-2232-INT 122-2231 123-2232-INT
AG122-2232-INTEA 928.53 USD InStock
AG122-2232-INT CAAG125-2237 AG123-2232-INT AG123-2232 AG125-2232 CAAG1232232INT CAAG1222232INT AG125-2212 AG122-2232 CA122-2232 AG122-2231 AG122-2212 CA122-2212 AG125-2237 CA123-2232 CAAG122-2231 CAAG125-2212 CAAG125-2232
J&W DB-225 Columns, Agilent Technologies
Chromatography Columns
Ideal for separations of cis- and trans-fatty acid methyl esters in food testing applications.
  • (50%-Cyanopropylphenyl)-dimethylpolysiloxane
  • Mid/high polarity
  • Excellent for separations of cis- and trans-fatty acid methyl esters (FAMEs)
  • Bonded, crosslinked, and solvent rinsable
  • Exact replacement of HP-225
  • Close equivalent to USP Phase G7
  • Tested with the tightest industry QC specifications for column bleed, sensitivity, and efficiency
  • Performance Summary shipped with each column
  • Similar phases: SP-2330, Rtx-225, BP-225, OV-225, 007-225, AT-225
Agilent J&W DB-225 provides excellent performance for food testing applications. These bonded columns are closely equivalent to USP phase G19 and can be used as exact replacements for HP-225 column types. DB-225 is crosslinked and solvent rinsable and is fully operational between temperature limits of 40 and 220/240 °C. These columns are excellent for effective separations of cis- and trans-fatty acid methyl esters.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty