Would you like to log in again?
CrossLab DB-2887 Columns, Agilent Technologies
Supplier: Agilent
CrossLab
125-2814 125-2814
AG125-2814EA 366.47 USD InStock
CA125-2814 AG125-2814
CrossLab DB-2887 Columns, Agilent Technologies
Chromatography Columns
Designed for the simulated distillation of ASTM D2887 with rapid conditioning, fast run time, and low bleed compared to packed columns.
  • 100% Dimethylpolysiloxane
  • Specifically designed for simulated distillation using ASTM Method D 2887
  • Rapid conditioning, fast run time, and low bleed compared to packed columns
  • Bonded, crosslinked, solvent rinsable
  • Tested with the tightest industry QC specifications for column bleed, sensitivity, and efficiency
  • Performance Summary shipped with each column
Agilent J&W DB-2887 is specifically designed for simulated distillation according to ASTM method D2887. With rapid conditioning, fast run times and low-bleed characteristics, these bonded, crosslinked, solvent rinsable columns deliver high column-to-column performance. A wide temperature operating range (–60 to 350 °C) and strict industry QC standards testing further enhance performance and usability.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty