Would you like to log in again?
DB-ALC1 and DB-ALC2 Columns, Agilent Technologies
Supplier: Agilent
J&W
123-9234 123-9234 125-9234 125-9234 125-9134 123-9134E 123-9134 125-9134 123-9134E 123-9134
AG123-9234EA 783.38 USD InStock
CAAG125-9234 AG123-9234 CA123-9234 CAAG123-9134E CA123-9134 AG123-9134 AG125-9234 AG123-9134E CA125-9134 AG125-9134
DB-ALC1 and DB-ALC2 Columns, Agilent Technologies
Chromatography Columns
Delivers blood alcohol analysis results in just 2 minutes, reducing run time by a third compared to other blood alcohol analysis columns.
  • Delivers reliable blood alcohol analysis
  • Optimized primary and confirmation column pair for US blood alcohol analysis
  • Faster GC run times
  • Improved resolution of key ethanol/acetone peaks
  • Available in 0.32 and 0.53 mm Int.Ø.
  • Bonded and crosslinked
  • Tested with the tightest industry QC specifications for column bleed, sensitivity, and efficiency
  • Performance Summary shipped with each column
Agilent J&W DB-ALC1 and DB-ALC2 analytical columns enable two-minute blood alcohol content analysis, reducing run time by one-third compared to other blood alcohol analysis columns. Because of its unique polarity, DB-ALC2 also provides better resolution of key ethanol/acetone peaks than other available blood alcohol confirmation columns.
These columns are suitable for both headspace analysis and direct injections.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty